1-(2-Hydroxy-5-(trifluoromethyl)phenyl)ethanone 97%

Supplier: Ambeed
Warning

A129696-250MG A129696-5G A129696-100MG A129696-1G
76811-112EA 42 USD
76811-112 76811-114 77883-039 77883-040
1-(2-Hydroxy-5-(trifluoromethyl)phenyl)ethanone 97%
1-(2-Hydroxy-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD11840324
CAS Number: 67589-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR