4-Propyl-2,3'',4'',5''-tetrafluoro-1,1':4',1''-terphenyl 97%

Supplier: Ambeed
Warning

A129324-1G A129324-5G
76811-096EA 6 USD
76811-096 76811-098
4-Propyl-2,3'',4'',5''-tetrafluoro-1,1':4',1''-terphenyl 97%
4-Propyl-2,3'',4'',5''-tetrafluoro-1,1':4',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₁H₁₆F₄
MDL Number: MFCD16038524
CAS Number: 205806-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR