N-Fmoc-trans-4-hydroxy-L-proline methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-trans-4-OH-Pro-OMe, (2S,4R)-1-((9H-Fluoren-9-yl)methyl) 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, Fmoc-Hyp-OMe

A128654-5G A128654-1G A128654-25G
76811-080EA 36 USD
76811-080 77611-132 77611-134
N-Fmoc-trans-4-hydroxy-L-proline methyl ester 97%
N-Fmoc-trans-4-hydroxy-L-proline methyl ester

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₅
MDL Number: MFCD12545917
CAS Number: 122350-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR