1-Bromo-3,5-dimethoxy-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A128610-100MG A128610-1G A128610-250MG
76811-072EA 9 USD
76811-072 76811-074 76811-076
1-Bromo-3,5-dimethoxy-2-nitrobenzene 97%
1-Bromo-3,5-dimethoxy-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₄
MDL Number: MFCD19442839
CAS Number: 1379358-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR