5-Methoxy-2-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-Methylphenylboronic acid, (5-Methoxy-2-methylphenyl)boronic acid

A128224-100MG A128224-1G A128224-250MG A128224-5G
76811-032EA 30 USD
76811-032 76811-034 76811-036 77883-035
5-Methoxy-2-methylphenylboronic acid 97%
5-Methoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD03425973
CAS Number: 617689-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR