3-Hydrazinylpyridine 95%

Supplier: Ambeed
Warning

A127791-100MG A127791-250MG
76811-010EA 43.5 USD
76811-010 76811-012
3-Hydrazinylpyridine 95%
3-Hydrazinylpyridine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD07786306
CAS Number: 42166-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR