4-Bromo-1-naphthaleneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromonaphthalen-1-yl)boronic acid

A127201-100MG A127201-10G A127201-1G A127201-250MG A127201-25G A127201-5G
76810-980EA 5.31 USD
76810-980 76810-982 76810-984 76810-986 76810-988 76810-990
4-Bromo-1-naphthaleneboronic acid 97%
4-Bromo-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BBrO₂
MW: 250.88 g/mol
MDL Number: MFCD08701764
CAS Number: 145965-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR