Ethyl 4-bromooxazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A127199-1G A127199-250MG A127199-25G A127199-5G
76810-972EA 132.62 USD
76810-972 76810-974 76810-976 76810-978
Ethyl 4-bromooxazole-5-carboxylate 97%
Ethyl 4-bromooxazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₃
MW: 220.02 g/mol
MDL Number: MFCD20260216
CAS Number: 1258283-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR