Methyl 6-methoxy-2-pyrazinecarboxylate 97%

Supplier: Ambeed
Warning

A126950-100MG A126950-250MG A126950-5G
76810-964EA 10 USD
76810-964 76810-966 77883-023
Methyl 6-methoxy-2-pyrazinecarboxylate 97%
Methyl 6-methoxy-2-pyrazinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD02258380
CAS Number: 23813-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR