2-Methyl-1H-pyrazol-3(2H)-one 97%

Supplier: Ambeed
Warning

A126502-1G A126502-250MG A126502-5G A126502-25G
76810-940EA 19 USD
76810-940 76810-942 76810-944 77883-021
2-Methyl-1H-pyrazol-3(2H)-one 97%
2-Methyl-1H-pyrazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MDL Number: MFCD20527389
CAS Number: 3310-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR