Methyl 5-hydroxy-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-hydroxy-1H-pyrazole-3-carboxylate

A126193-100MG A126193-1G A126193-250MG A126193-5G
76810-922EA 12.74 USD
76810-922 76810-924 76810-926 76810-928
Methyl 5-hydroxy-pyrazole-3-carboxylate 95%
Methyl 5-hydroxy-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MW: 142.113 g/mol
MDL Number: MFCD09878841
CAS Number: 1018446-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR