3-Bromo-2-chloro-6-fluorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A126116-1G
A126116-250MG
A126116-25G
A126116-5G
76810-914EA
40
USD
76810-914
76810-916
76810-918
76810-920
3-Bromo-2-chloro-6-fluorobenzaldehyde 98%
3-Bromo-2-chloro-6-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrClFO |
MDL Number:
MFCD11110260 CAS Number: 1114809-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...