Ethyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A126035-100MG A126035-250MG
76810-910EA 24 USD
76810-910 76810-912
Ethyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%
Ethyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O₂
MDL Number: MFCD17168482
CAS Number: 1269293-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR