4-(4-Chlorophenyl)cyclohexanol 95%

Supplier: Ambeed
Warning

A125786-100MG A125786-1G A125786-250MG
76810-900EA 56 USD
76810-900 76810-902 76810-904
4-(4-Chlorophenyl)cyclohexanol 95%
4-(4-Chlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO
MDL Number: MFCD11850098
CAS Number: 930766-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR