3-Bromo-6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one 98%

Supplier: Ambeed
Warning

A125755-100MG A125755-250MG
76810-896EA 41 USD
76810-896 76810-898
3-Bromo-6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one 98%
3-Bromo-6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO
MDL Number: MFCD11036254
CAS Number: 87779-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR