Methyl 2-(4,6-dichloropyrimidin-5-yl)acetate 97%
Supplier: Ambeed
|
Warning
|
A125624-100MG
A125624-10G
A125624-250MG
A125624-5G
A125624-1G
76810-888EA
10.61
USD
76810-888
76810-890
76810-892
76810-894
77670-782
Methyl 2-(4,6-dichloropyrimidin-5-yl)acetate 97%
Methyl 2-(4,6-dichloropyrimidin-5-yl)acetate
Precaución: for Research Use Only
Formula:
C₇H₆Cl₂N₂O₂ MW: 221.04 g/mol |
MDL Number:
MFCD20526414 CAS Number: 171096-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...