4-Amino-3-bromobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-bromobenzoic Acid

A125578-100G A125578-10G A125578-1G A125578-25G A125578-500G A125578-5G
76810-876EA 188.85 USD
76810-876 76810-878 76810-880 76810-882 76810-884 76810-886
4-Amino-3-bromobenzoic acid 98%
4-Amino-3-bromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
MDL Number: MFCD03407439
CAS Number: 6311-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR