5-(Benzyloxy)-2-bromobenzaldehyde 97%

Supplier: Ambeed
Warning

A125341-100MG A125341-1G A125341-250MG A125341-5G
76810-838EA 5 USD
76810-838 76810-840 76810-842 76810-844
5-(Benzyloxy)-2-bromobenzaldehyde 97%
5-(Benzyloxy)-2-bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
MDL Number: MFCD03425931
CAS Number: 85604-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR