tert-Butyl 4-(cyclopropanecarbonyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A124826-100MG A124826-5G A124826-1G
76810-774EA 13.8 USD
76810-774 76810-776 77670-770
tert-Butyl 4-(cyclopropanecarbonyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(cyclopropanecarbonyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₃
MW: 254.32 g/mol
MDL Number: MFCD03930967
CAS Number: 414910-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR