4-Bromo-5-methyl-2-nitrophenol 98%

Supplier: Ambeed
Danger

A124566-100MG A124566-1G A124566-250MG
76810-752EA 13 USD
76810-752 76810-754 76810-756
4-Bromo-5-methyl-2-nitrophenol 98%
4-Bromo-5-methyl-2-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD12923347
CAS Number: 182500-28-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR