4-Bromo-1-(tert-butyl)-5-phenyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

A124550-100MG A124550-250MG
76810-748EA 8 USD
76810-748 76810-750
4-Bromo-1-(tert-butyl)-5-phenyl-1H-pyrazole 95%
4-Bromo-1-(tert-butyl)-5-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂
MW: 279.18 g/mol
MDL Number: MFCD19704856
CAS Number: 1269291-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR