Diethyl-4-oxocyclohexane-1,1-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Diethyl 4-oxocyclohexane-1,1-dicarboxylate

A124522-100MG A124522-10G A124522-1G A124522-250MG A124522-25G A124522-5G
76810-726EA 10.61 USD
76810-726 76810-728 76810-730 76810-732 76810-734 76810-736
Diethyl-4-oxocyclohexane-1,1-dicarboxylate 95%
Diethyl-4-oxocyclohexane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₅
MW: 242.27 g/mol
MDL Number: MFCD00177942
CAS Number: 55704-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR