4-Nitro-1-isoindolinone 95%

Supplier: Ambeed
Warning

Synonyms: 4-Nitroisoindolin-1-one

A124306-100MG A124306-1G A124306-250MG
76810-714EA 11 USD
76810-714 76810-716 76810-718
4-Nitro-1-isoindolinone 95%
4-Nitro-1-isoindolinone

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD09701290
CAS Number: 366452-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR