tert-Butyl exo-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate 98%

Supplier: Ambeed
Warning

A123720-10G A123720-1G A123720-25G A123720-5G A123720-250MG
76810-674EA 228.1 USD
76810-674 76810-676 76810-678 76810-680 77592-414
tert-Butyl exo-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl exo-3-amino-8-azabicyclo[3.2.1]octane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.31 g/mol
MDL Number: MFCD18428076
CAS Number: 744183-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR