Fmoc-Ile-ol 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-isoleucinol

A123710-1G A123710-5G
76810-670EA 13 USD
76810-670 76810-672
Fmoc-Ile-ol 97%
Fmoc-Ile-ol

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₃
MW: 339.40 g/mol
MDL Number: MFCD00235930
CAS Number: 133565-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR