1H-Imidazole-2-carboxamide 97%

Supplier: Ambeed
Warning

A123671-100MG A123671-1G A123671-250MG A123671-5G
76810-660EA 20 USD
76810-660 76810-662 76810-664 76810-666
1H-Imidazole-2-carboxamide 97%
1H-Imidazole-2-carboxamide

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD09261275
CAS Number: 16093-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR