4-Methylbenzo[d]thiazol-6-ol 95%

Supplier: Ambeed
Warning

A123373-100MG A123373-1G A123373-250MG
76810-648EA 120.95 USD
76810-648 76810-650 76810-652
4-Methylbenzo[d]thiazol-6-ol 95%
4-Methylbenzo[d]thiazol-6-ol

Precaución: for Research Use Only

Formula: C₈H₇NOS
MW: 165.21 g/mol
MDL Number: MFCD12962873
CAS Number: 1190317-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR