4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine maleate 98+%

Supplier: Ambeed
Warning

Synonyms: RS-127445 maleate

A123169-100MG A123169-250MG A123169-1G
76810-636EA 142 USD
76810-636 76810-638 77670-748
4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine maleate 98+%
4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine maleate

Precaución: for Research Use Only

Formula: C₂₁H₂₀FN₃O₄
MW: 397.39 g/mol
MDL Number: MFCD21337892
CAS Number: 199864-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR