1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile 95%
Supplier: Ambeed
|
Warning
|
A123098-100MG
A123098-250MG
A123098-1G
76810-632EA
118.83
USD
76810-632
76810-634
77670-744
1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile 95%
1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile
Precaución: for Research Use Only
Formula:
C₁₆H₂₀BNO₂ MW: 269.15 g/mol |
MDL Number:
MFCD19689506 CAS Number: 1346264-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...