Ethyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A122629-1G A122629-250MG
76810-586EA 82.76 USD
76810-586 76810-588
Ethyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate 95%
Ethyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O₂S
MW: 277.14 g/mol
MDL Number: MFCD18642497
CAS Number: 74840-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR