5-Bromo-4-chloro-2-methylpyridine 95%

Supplier: Ambeed
Warning

A122422-250MG A122422-1G
76810-562EA 19 USD
76810-562 77670-736
5-Bromo-4-chloro-2-methylpyridine 95%
5-Bromo-4-chloro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MDL Number: MFCD09839296
CAS Number: 1003711-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR