2-Chloro-6-iodobenzoic acid 97%

Supplier: Ambeed
Warning

A122391-1G A122391-250MG A122391-5G A122391-10G
76810-558EA 15 USD
76810-558 76810-560 77571-360 77882-987
2-Chloro-6-iodobenzoic acid 97%
2-Chloro-6-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
MDL Number: MFCD06808823
CAS Number: 13420-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR