2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A122117-100MG
A122117-1G
A122117-250MG
76810-528EA
26.53
USD
76810-528
76810-530
76810-532
2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₇H₆ClN₃ MW: 167.596 g/mol |
MDL Number:
MFCD13189420 CAS Number: 1060816-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...