8-(Trifluoromethyl)chroman-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

A122046-100MG A122046-250MG
76810-524EA 31 USD
76810-524 76810-526
8-(Trifluoromethyl)chroman-4-amine hydrochloride 95%
8-(Trifluoromethyl)chroman-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClF₃NO
MW: 253.65 g/mol
MDL Number: MFCD08544298
CAS Number: 1392212-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR