3-Chloro-5-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A121893-100G A121893-10G A121893-1G A121893-250MG A121893-25G A121893-5G
76810-512EA 328 USD
76810-512 76810-514 76810-516 76810-518 76810-520 76810-522
3-Chloro-5-methylbenzoic acid 97%
3-Chloro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD11007823
CAS Number: 56961-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR