4-Methoxy-2,6-dimethylaniline 97%

Supplier: Ambeed
Warning

A121438-100MG A121438-1G A121438-250MG A121438-5G
76810-494EA 14 USD
76810-494 76810-496 76810-498 77566-250
4-Methoxy-2,6-dimethylaniline 97%
4-Methoxy-2,6-dimethylaniline

Precaución: For research use only.

Formula: C₉H₁₃NO
MDL Number: MFCD03792670
CAS Number: 34743-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR