5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine dihydrochloride 95%

Supplier: Ambeed
Warning

A120996-1G A120996-5G
76810-492EA 44 USD
76810-492 77670-714
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine dihydrochloride 95%
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁Cl₂N₃
MW: 196.08 g/mol
MDL Number: MFCD09954907
CAS Number: 165894-10-0
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR