Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate 97%

Supplier: Ambeed
Warning

Synonyms: Amfenac Sodium Hydrate, Amfenac sodium, Sodium 2-(2-amino-3-benzoylphenyl)acetate hydrate, Fenazox, Amfenac Sodium Monohydrate

A120991-10MG A120991-1G A120991-5G A120991-25G A120991-50MG
76810-486EA 6 USD
76810-486 76810-488 76810-490 77882-980 77882-981
Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate 97%
Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate

Precaución: for Research Use Only

Formula: C₁₅H₁₄NNaO₄
MW: 295.2657 g/mol
MDL Number: MFCD00941383
CAS Number: 61618-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR