trans,trans-Dibenzylideneacetone 98%

Supplier: Ambeed
Warning

Synonyms: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

A120658-100G A120658-10G A120658-1G A120658-25G A120658-5G A120658-500G
76810-476EA 48 USD
76810-476 76810-478 76810-480 76810-482 76810-484 77571-228
trans,trans-Dibenzylideneacetone 98%
trans,trans-Dibenzylideneacetone

Precaución: for Research Use Only

Formula: C₁₇H₁₄O
MW: 234.30 g/mol
MDL Number: MFCD00004790
CAS Number: 35225-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR