Adenosine-5'-diphosphoric acid (ADP) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Adenosine 5'-diphosphate, 5′-ADP, Adenosine 5′-diphosphate
A120434-1G
A120434-5G
A120434-25G
76810-474EA
16.98
USD
76810-474
77611-020
77670-710
Adenosine-5'-diphosphoric acid (ADP) 95%
Adenosine-5'-diphosphoric acid (ADP)
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N₅O₁₀P₂ MW: 427.2 g/mol |
MDL Number:
MFCD00066473 CAS Number: 58-64-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...