4-Fluorobenzo[b]thiophene 95%

Supplier: Ambeed
Warning

A120417-100MG A120417-1G A120417-250MG
76810-468EA 46.68 USD
76810-468 76810-470 76810-472
4-Fluorobenzo[b]thiophene 95%
4-Fluorobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅FS
MW: 152.18 g/mol
MDL Number: MFCD16657500
CAS Number: 310466-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR