2-(Piperidin-3-yl)acetic acid hydrochloride 95%

Supplier: Ambeed
Warning

A120360-1G A120360-250MG A120360-25G A120360-5G
76810-460EA 22.28 USD
76810-460 76810-462 76810-464 76810-466
2-(Piperidin-3-yl)acetic acid hydrochloride 95%
2-(Piperidin-3-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.6446 g/mol
MDL Number: MFCD02181158
CAS Number: 71985-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR