3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid 95%

Supplier: Ambeed
Warning

A120310-100MG A120310-1G A120310-250MG A120310-5G
76810-452EA 90.18 USD
76810-452 76810-454 76810-456 76810-458
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid 95%
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇F₃O₃
MW: 184.12 g/mol
MDL Number: MFCD11870383
CAS Number: 1163729-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR