3-(4-Bromophenyl)-9-phenylcarbazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Bromophenyl)-9-phenyl-9H-carbazole

A120292-1G A120292-25G A120292-5G
76810-446EA 18.04 USD
76810-446 76810-448 76810-450
3-(4-Bromophenyl)-9-phenylcarbazole 98%
3-(4-Bromophenyl)-9-phenylcarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
MDL Number: MFCD14582939
CAS Number: 1028647-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR