Ethyl-2-methyl-1,3-oxazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methyloxazole-4-carboxylate

A120204-100MG A120204-10G A120204-1G A120204-250MG A120204-25G A120204-5G
76810-434EA 14.86 USD
76810-434 76810-436 76810-438 76810-440 76810-442 76810-444
Ethyl-2-methyl-1,3-oxazole-4-carboxylate 98%
Ethyl-2-methyl-1,3-oxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD07367959
CAS Number: 10200-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR