Methyl 2-amino-5-nitronicotinate 97%

Supplier: Ambeed
Warning

A119888-250MG A119888-5G
76810-416EA 16.98 USD
76810-416 76810-418
Methyl 2-amino-5-nitronicotinate 97%
Methyl 2-amino-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₇H₇N₃O₄
MW: 197.15 g/mol
MDL Number: MFCD19441225
CAS Number: 88312-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR