4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride
A119392-10G
A119392-1G
A119392-250MG
A119392-25G
A119392-5G
76810-380EA
220.67
USD
76810-380
76810-382
76810-384
76810-386
77611-002
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 98%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₉N₃·HCl MW: 159.62 g/mol |
MDL Number:
MFCD09999261 CAS Number: 879668-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...