[1,2,4]Triazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A118682-1G A118682-250MG
76810-338EA 35 USD
76810-338 77882-968
[1,2,4]Triazolo[1,5-a]pyridine 97%
[1,2,4]Triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD03700708
CAS Number: 274-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR