4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,4-Trifluoro-1-(p-tolyl)butane-1,3-dione, 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione

A118278-100G A118278-10G A118278-25G A118278-500G A118278-5G
76810-308EA 78.51 USD
76810-308 76810-310 76810-312 76810-314 76810-316
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione 97%
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₂
MW: 230.19 g/mol
MDL Number: MFCD00517909
CAS Number: 720-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR