Methyl-2-oxotetrahydrofuran-3-carboxylate 95%

Supplier: Ambeed
Warning

A118145-1G A118145-250MG A118145-100MG A118145-5G
76810-292EA 47 USD
76810-292 76810-294 77882-965 77882-966
Methyl-2-oxotetrahydrofuran-3-carboxylate 95%
Methyl-2-oxotetrahydrofuran-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MDL Number: MFCD10569624
CAS Number: 19406-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR